CytoVista™ 3D Cell Culture Clearing Reagent - FAQs
CytoVista™ 3D Cell Culture Clearing Reagent - FAQs
View additional product information for CytoVista™ 3D Cell Culture Clearing Reagent - FAQs (V11316)
1 product FAQs found
Do I have to use CytoVista 3D Cell Culture Clearing Reagent under a chemical safety hood?
We recommend following the usage guidelines specified in the MSDS.