CytoVista™ Tissue Imaging Chamber, 1.75 mm deep - FAQs
CytoVista™ Tissue Imaging Chamber, 1.75 mm deep - FAQs
View additional product information for CytoVista™ Tissue Imaging Chamber, 1.75 mm deep - FAQs (V11317)
3 product FAQs found
Do I need to seal the outer edges of the CytoVista Tissue Imaging chamber touching the glass, to prevent liquid from leaking out?
This should not be required as the silicone naturally adheres to the glass. If sealing is desired, you may seal with melted paraffin or VALAP at the outer edges.
What are the dimensions of the CytoVista Tissue Imaging chambers?
The CytoVista Tissue Imaging chambers have an outer perimeter of 25 mm x 25 mm and an inner diameter of 16 mm; they vary in thickness.
How can I paste the CytoVista Tissue Imaging chamber onto a glass coverslip or slide?
The silicone naturally adheres to glass.