Can spheroids or organoids be imaged and stored in CytoVista 3D Cell Culture Clearing Reagents (Cat. Nos. V11315, V11326)?
CytoVista 3D Cell Culture Clearing Reagent (Cat. Nos. V11315, V11326) can be used as an imaging solution. However, we do not recommend keeping the spheroids or organoids in the clearing reagent for more than two weeks.