Image-iT™ Plant Tissue Hard-Set Mountant, 30 mL - FAQs
Image-iT™ Plant Tissue Hard-Set Mountant, 30 mL - FAQs
View additional product information for Image-iT™ Plant Tissue Hard-Set Mountant - FAQs (V11331)
4 product FAQs found
Can I remove Image-iT Plant Tissue Hard-Set Mountant from my sample once cured?
Once cured, Image-iT Plant Tissue Hard-Set Mountant cannot be removed from the sample.
I accidentally froze Image-iT Plant Tissue Hard-Set Mountant. Can I still use it?
We do not recommend using Image-iT Plant Tissue Hard-Set Mountant that has been frozen.
How should I store samples mounted in Image-iT Plant Tissue Hard-Set Mountant?
We recommend storing the slides at room temperature in a moisture-free environment.
How long can I store samples mounted in Image-iT Plant Tissue Hard-Set Mountant?
Slides can be mounted permanently by drying excess Image-iT Plant Tissue Clearing Reagent with a lab tissue and applying Image-iT Plant Tissue Hard-Set Mountant.