CytoVista™ Sagittal Mouse Brain Slicer, 1 mm - FAQs
CytoVista™ Sagittal Mouse Brain Slicer, 1 mm - FAQs
View additional product information for CytoVista™ Sagittal Mouse Brain Slicer, 1 mm - FAQs (V11334)
3 product FAQs found
Can I autoclave the CytoVista Sagittal Mouse Brain Slicer, 1 mm?
No, the CytoVista Sagittal Mouse Brain Slicer, 1 mm cannot be autoclaved.
How can I clean the CytoVista Sagittal Mouse Brain Slicer, 1 mm?
Clean with soap/detergent and cold water, rinse thoroughly and dry completely after cleaning. Expose to water only long enough to clean the slicer; do not soak in water for prolonged periods. Sonication and heat may distort the slicer.
What type of material is the CytoVista Sagittal Mouse Brain Slicer, 1 mm made of?
It is composed of PLA (polylactic acid).