Sensititre AIM™ Automated Inoculation Delivery System
Sensititre AIM™ Automated Inoculation Delivery System
Sensititre AIM™ Automated Inoculation Delivery System
Sensititre AIM™ Automated Inoculation Delivery System
Thermo Scientific™

Sensititre AIM™ Automated Inoculation Delivery System

Quickly and accurately dose most 96-well microtitre plates, eliminating both skipped wells and costly repeat tests.
Have Questions?
Catalog NumberHeight (English)
V302011.3 in.
Catalog number V3020
Price (USD)
-
Height (English):
11.3 in.
Quickly and accurately dose most 96-well microtitre plates, eliminating both skipped wells and costly repeat tests, with the Thermo Scientific™ Sensititre AIM™ Automated Inoculation Delivery System. Just select your dosing volume and pattern with the easy-to-use, icon-driven touch screen for simple, intuitive plate inoculation. With its compact, user-friendly design, the AIM System also provides flexible storage and accessibility with minimal benchtop footprint to conserve valuable laboratory space.
  • Compatibility with most 96-well microtitre plates ensures flexibility with dry or frozen susceptibility formats
  • Unique tube design and manually applied seal provide additional protection against biohazardous spill concerns
Not all products are available for sale in all territories. Please inquire.

VersaTREK™ and Sensititre products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

Specifications
Depth (English)12.3 in.
Depth (Metric)31.2 cm
DescriptionSensititre AIM Automated Inoculation Delivery System
Electrical Requirements94-237V; 47-63Hz; 150w
Height (English)11.3 in.
Height (Metric)28.7 cm
Regulatory StatusFDA-cleared
Weight (English)17.6 lb.
Weight (Metric)8 kg
Width (English)13.3 in.
Width (Metric)33.8 cm
Product TypeMicrobiology Media
Test TypeMinimum Inhibitory Concentration (MIC)
Unit SizeEach