Vybrant™ FLICA Caspase Apoptosis Assay Kits for flow cytometry, poly caspases - FAQs
Vybrant™ FLICA Caspase Apoptosis Assay Kits for flow cytometry, poly caspases - FAQs
View additional product information for Vybrant™ FLICA Caspase Apoptosis Assay Kits for flow cytometry - FAQs (V35117, V35118, V35119)
1 product FAQs found
What are the fluorescence excitation/emission maxima for the FAM-VAD-FMK FLICA reagent, Hoechst 33342, and propidium iodide supplied in the Vybrant FAM Poly Caspases Assay Kit, for flow cytometry?
FAM-VAD-FMK FLICA reagent: 488/530 nm
Hoechst 33342: 350/461 nm
Propidium iodide: 535/617 nm