Owl™ VEP-2 Mini Tank Electroblotting System
Thermo Scientific™

Owl™ VEP-2 Mini Tank Electroblotting System

Produce uniform, reproducible protein transfers for up to four polyacrylamide minigels with the Thermo Scientific™ Owl™ VEP-2 Mini Tank Electroblotting System.
Have Questions?
Catalog NumberType
VEP-2Electroblotting System
Catalog number VEP-2
Price (USD)
1,468.83
Online exclusive
1,690.00
Save 221.17
Each
Type:
Electroblotting System
Price (USD)
1,468.83
Online exclusive
1,690.00
Save 221.17
Each

Produce uniform, reproducible protein transfers for up to four polyacrylamide minigels with the Thermo Scientific™ Owl™ VEP-2 Mini Tank Electroblotting System, ideal for fast, efficient transfer of proteins over a wide molecular weight range.

Features:
  • Tank-style electroblotting system
  • Integrated cooling base enables temperature-controlled runs
  • Stir bars (not included) may be added to bottom of buffer chamber for increased buffer circulation

Includes:

  • Buffer chamber with platinum wire electrode panels and cooling base
  • Safety interlocking lid with attached power supply leads
  • Four blotting cassettes with foam pads and cassette clips
  • Six sheets of blotting filter paper
Specifications
Dimensions (L x W x H)7.06 x 5.13 x 6.5 in. (18 x 13 x 16.5 cm)
Height (English)6.5 in.
Height (Metric)16.5 cm
Length (English)7.06 in.
Length (English) Gel3.5 in.
Length (Metric)18 cm
Length (Metric) Gel9 cm
Volume (English) Buffer0.3 gal.
Volume (Metric) Buffer1.4 L
Width (English)5.13 in.
Width (English) Gel3.5 in.
Width (Metric)13 cm
Width (Metric) Gel9 cm
Gel SizeMini (9 in. x 9 in.)
Product LineOwl
TypeElectroblotting System
Unit SizeEach