Can checkweighers work with pharmaceutical products?
The Thermo Scientific Versa Rx and Versa RxV Pharmaceutical Checkweighers (https://www.thermofisher.com/order/catalog/product/VERSARXPHARMA) are specifically designed for pharmaceutical applications. These systems are highly accurate - within ±50 mg at 3 sigma and speeds up to 550 ppm. They have a clean design with a slack belt knife edge infeed transfer for unparalleled accuracy and optimal product handling. They are ideal for a variety of products, including blister packs, cartons, plastic bottles, glass bottles, pouches, and small aerosols.