Vanquish™ Charged Aerosol Detectors
Thermo Scientific™

Vanquish™ Charged Aerosol Detectors

Performance you can see. Confidently detect and quantify all non-volatile and many semi-volatile analytes with Thermo Scientific™ Vanquish™ Charged Aerosol Detectors.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberModelTemperature Range
VH-D20-AVanquish CAD HAmbient +5° to +100°C
VH-D21-A-01Vanquish CAD HPAmbient, 10 to 100°C
VF-D21-A-01Vanquish CAD FPAmbient, 35°C, 50°C, 70°C
6085.0307Vanquish CAD HP, Analog OutputAmbient, 10°C to 100°C
6085.0310Vanquish CAD HP, Digital OutputAmbient, 10°C to 100°C
Catalog number VH-D20-A
Price (TWD)
-
Model:
Vanquish CAD H
Temperature Range:
Ambient +5° to +100°C

Expand your detection range and get accurate quantitation in your high-performance liquid chromatography (HPLC) analysis with the near-universal Charged Aerosol Detector (CAD). Confidently quantify non-volatile and many semi-volatile analytes. Thermo Scientific™ Vanquish™ CADs deliver sensitive and consistent response across compound classes, helping you accurately measure both low-level impurities and highly concentrated compounds in a single analysis.

Why choose a Vanquish™ Charged Aerosol Detector?

Confidently quantify non-volatile and many semi-volatile analytes. Vanquish CAD systems combine near-universal detection, reliable single-calibrant quantitation of unknowns, superior sensitivity versus ELSD, and compatibility with gradient methods. Simplify quantitation, from trace impurities to highly concentrated compounds.

Designed around your workflow

The Vanquish™ CAD P Series improves reproducibility, simplifies method transfer, supports compliance, and integrates with your preferred CDS platform.

 Improve quantitation accuracy and reduce contamination risk

  • Integrated diverter valve for enhanced flow-path control

 Increase method robustness and reproducibility for semi-volatile analytes

  • Temperature coupling mode in the charging detection module

Accelerate method development and optimize detector response

  • Four simultaneous power levels and support for inverse gradient workflows

Simplify method transfer between CAD generations

  • Built-in method translation tools in Chromeleon™ CDS

Continue using your preferred CDS and established workflows   

  • Compatible with Chromeleon™ CDS control and Thermo Scientific™ SII integration with Empower™ and OpenLab CDS; compatible with virtually any LC system

Support compliant operation and simplify qualification

  • Audited flow-path control, qualification, and functional testing tools

Product offerings

Vanquish™ CAD P Series

  • Vanquish™ CAD HP
  • Vanquish™ CAD FP

Integration with other HPLC/UHPLC instruments

  • Digital and Analog Output options available

 Legacy Vanquish™ CAD

  • Vanquish™ CAD H

 

Specifications
Communications1 USB port (USB 2.0, “B” type) 1 USB hub with 3 ports (USB 2.0, “A” type) to connect further Vanquish modules
Detection ModeRecords a single power value
For Use With (Equipment)Vanquish HPLC/UHPLC systems
GLP FeaturesPredictive Performance functions: Gas filter change interval, service monitoring period, system parameters logged in the Chromeleon Chromatography Data System Audit Trail.
Gas RequirementsNitrogen (recommended) or Compressed Air
ModelVanquish CAD H
RangeDynamic Range: Up to 4 orders of magnitude
Safety FeaturesPower-up diagnostics, inlet gas over-pressure, high evaporation temp, pump flow shutoff, fluid leak detection and safe leak handling
Temperature ControlSettable
Temperature RangeAmbient +5° to +100°C
Data Collection Rate200 Hz Max.
TypeStandard flow management
Unit SizeEach