Vanquish™ Amplify UHPLC systems
Thermo Scientific™

Vanquish™ Amplify UHPLC systems

Green features

Expanding the Thermo Scientific™ Vanquish™ LC portfolio with a fully inert sample flow path ideal for metal-sensitive molecules and preventing metals adducts in LC-MS applications.

Have Questions?
Catalog NumberType
VQ-AMPLIFYIntegrated Inert System
Catalog number VQ-AMPLIFY
Price (USD)
-
Type:
Integrated Inert System

Designed for demanding biopharmaceutical workflows, including oligonucleotide, mRNA, protein therapeutics, and antibody-drug conjugate analysis, the system's inert sample path helps prevent unwanted interactions between analytes and metal surfaces, reducing adsorption and supporting improved recovery, reproducibility, and confidence in chromatographic results; run after run, what you see isn't just what you get. It's what you can depend on. Vanquish™ Amplify UHPLC systems integrate with Thermo Scientific™ SurePac™ RP MDi columns, the Thermo Scientific™ OrbitrapTM Excedion™ mass spectrometer, and Thermo Scientific™ Chromeleon™ CDS software to support LC–MS workflows, compliant data management, and dependable results from early research through manufacturing QC.

The Vanquish Amplify UHPLC Systems are based on the Vanquish UHPLC systems platform featuring:

Fully inert and biocompatible sample flow path

Minimize non-specific adsorption of metal-sensitive analytes such as oligonucleotides and RNA, improving recovery, peak shape, reproducibility, and reducing metal adduct formation.

Thermo Scientific™ Viper™ TQ UHPLC Fittings

Secure & leak-free fluidic connections through a built-in torque control mechanism ensuring consistent and easy installation that you can hear and feel, preventing over- and under-tightened connections.

High-pressure UHPLC capability

Supports demanding high-resolution separations for oligonucleotides and nucleic acids on modern UHPLC methods.

Vial bottom detection

Supports maximized sample volume utilization by enabling the sample draw right from the bottom of the sample container and improved workflow reliability by removing the uncertainty of which settings to use, preventing needle damage.

Specifications
Flow Accuracy±0.1%
Green FeaturesACT Ecolabel (Platinum)
Max. Detector Channels10 + 3 D field
Sample Temperature Range4°C to 40°C (=22 K below ambient (at <80% RH))
TypeIntegrated Inert System
Column Temperature Range5°C to 120°C
Detector Data Collection Rate200 Hz Max.
Max. Pressure150 MPa
Solvent Channels2 x 3
Unit SizeEach