Vanquish™ Method Development HPLC and UHPLC System
Vanquish™ Method Development HPLC and UHPLC System
Thermo Scientific™
Vanquish™ Method Development HPLC and UHPLC System
Save time by accelerating and automating your HPLC and UHPLC method development process with Thermo Scientific™ Vanquish™ Method Development systems.
Have Questions?
Catalog number VQ-METHOD-DEV
Price (MXN)
-
Request A Quote
Accelerate and automate your method development using Thermo Scientific™ Vanquish Method Development HPLC and UHPLC systems. These systems can optimize separation conditions by testing different combinations of buffers on up to six HPLC columns using Thermo Scientific™ Chromeleon™ CDS eWorkflows. Achieve fully automated method development with ChromSwordAuto Chromeleon Connect and ChromSword™ AutoRobust Chromeleon Connect. Chromeleon CDS is also compatible with the S-Matrix™ Fusion QbD™ software packages.
View Vanquish Method Development HPLC resourcesHPLC Support Center
Simplified method setup
  • Chromeleon CDS features downloadable eWorkflows to simplify setup for automated column and buffer scouting
  • Chromeleon Data Vault enables secure and central data storage
Extensive, automated column and eluent screening option
  • When equipped with a solvent selection valve, system can perform extensive, automated column and eluent screening
  • The Thermo Scientific™ Viper™ Method Scouting Kit includes all the tubing and near-zero dead volume connections to screen up to four columns
Compatible software integrates with Chromeleon CDS for accelerated, automated method development
  • The Vanquish Method Development HPLC and UHPLC systems are controlled by Chromeleon CDS and compatible with leading method development software packages, such as ChromSwordAuto Chromeleon Connect or Fusion QbD.
  • ChromSword AutoRobust Chromeleon Connect automates design and execution of robustness testing. The software allows up to nine variables to be easily evaluated following design of experiments (DOE) principles.
  • Chromeleon ICH method validation templates can accelerate method validation.
ChromSword is a trademark of VWR International BVBA. S-Matrix and Fusion QbD are trademarks of S-Matrix Corporation.
Specifications
DescriptionVanquish Method Development HPLC and UHPLC systems
For Use With (Application)Automate method scouting, develop and optimize methods, test method robustness and accelerate method validation
For Use With (Equipment)Vanquish Horizon, Vanquish Flex or Vanquish Core Components
Max. Pressure150 MPa, depending on configuration
TypeIntegrated HPLC or Biocompatible UHPLC system
Unit SizeEach