Orion™ Versa Star Pro™ pH/ISE/Conductivity/Dissolved Oxygen Multiparameter Benchtop Meter
Thermo Scientific™

Orion™ Versa Star Pro™ pH/ISE/Conductivity/Dissolved Oxygen Multiparameter Benchtop Meter

Get premium performance and flexibility with our multiparameter benchtop meter with pH/ISE, conductivity and RDO/DO modules to measure up to four channels at once.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct Type
VSTAR92Meter; pH/Ion, Conductivity and DO; Standard Kit
VSTAR90Meter; pH/Ion, Conductivity and DO (No Sensor)
VSTAR94Meter; pH/Ion, Conductivity and DO; Optical DO Kit
VSTAR91Meter; 2 Ch. pH/Ion, Cond. and DO (No Sensor)
VSTAR93Meter; 2 Ch. pH/Ion, Cond. and DO; Standard Kit
Catalog number VSTAR92
Price (USD)
4,743.00
Online exclusive
5,270.00
Save 527.00
Each
Product Type:
Meter; pH/Ion, Conductivity and DO; Standard Kit
Price (USD)
4,743.00
Online exclusive
5,270.00
Save 527.00
Each

Get high accuracy, premium performance and ultimate flexibility with the Thermo Scientific™ Orion™ Versa Star Pro™ pH/ISE/Conductivity/Dissolved Oxygen Multiparameter Benchtop Meter. Meet your most challenging applications for pH, ion concentration, conductivity, dissolved oxygen and temperature. Easily view measurements on the stunning color display and customize the four meter channels with interchangeable modules. Log up to 2000 data point sets with time/date stamp and easily transfer logs via USB or RS232 to a printer or computer. Select systems with one or two pH/ISE modules.

Thermo Scientific Orion Versa Star Pro Benchtop Meters are designed to meet your most challenging application with ease. Delivering rich functionality and features, you can optimally configure your meter system to meet changing requirements. The Orion Versa Star Pro Benchtop Meters offer interchangeable measurement modules that allow multiple users to customize four separate channels to meet their specific requirements. Easily view your measurements on a stunning color display with adjustable text sizes. Take the guesswork out of meter calibration and setup with onscreen instructions available in a variety of local languages. Enjoy quick and simple data entry using the numeric keypad.

  • Onscreen measurement stability indicator lets you know exactly when your measurements are ready
  • Selectable read modes include Auto-Read to hold a stable reading, Continuous to show changing readings, Timed to log data at specific intervals and Single-Shot to hold a reading after a specific wait time is reached
  • Audible alarms automatically notify you when a set offset value, high/low limit value and calibration time interval are reached
  • Uniformly mix solutions using up to two meter-controlled stirrer probes that are easy to position and can be quickly rinsed between samples
  • Data log collects and date/time stamps up to 2000 measurement sets with optional electrode ID, sample ID and account ID
  • Calibration log stores 30 most recent calibrations per parameter with viewable graph for multipoint pH and ISE calibrations
  • Ensure measurement integrity by viewing the active calibration log while taking readings
  • Non-volatile meter memory preserves data and settings, even in the event of power loss
  • Meter facilitates data transfers to a computer via USB or RS232 connections
  • Meters include CE, TUV 3-in-1 and FCC Class A certifications
  • 3-year replacement warranty

Includes Orion Versa Star pH/ISE Measurement Module:

  • Measure pH, ion concentration, mV, relative mV or ORP with temperature
  • Perform up to a 6 point pH calibration using automatic buffer recognition of NIST, DIN or user-defined buffer groups
  • Perform up to a 6 point ISE calibration with selectable units of ppm, M, mg/L, %, ppb or none

Includes Orion Versa Star Conductivity Measurement Module:

  • Measure conductivity, TDS, salinity or resistivity with temperature
  • Perform up to a 6 point conductivity calibration
  • Use linear, non-linear, EP (USP) or off conductivity temperature compensation with 5°C, 10°C, 15°C, 20°C or 25°C reference temperatures

Includes Orion Versa Star RDO/DO Measurement Module:

  • Measure dissolved oxygen in percent saturation or mg/L with temperature using Orion RDO optical DO probes or Orion polarographic DO probes
  • Calibrate DO using water-saturated air, air-saturated water, custom value (Winkler titration) and zero point options
  • Automatically compensate DO readings using ATC temperature, manual salinity input and built-in barometer for barometric pressure

System Access Feature
Orion Versa Star Pro Meters offer a system access feature that is designed to help ensure data integrity by preventing undesired access to various meter functions including date/time setting, data log and calibration log. The System Access feature can be easily turned on or off as the needs of your lab change for ultimate flexibility in operation and ease-of-use. When enabled, the System Access feature allows assignment of a system administrator and up to 10 user accounts with customizable access to instrument settings, measurement settings, calibrations and logs. Use the password protection to secure selected meter functions, while leaving other functions accessible for all users. The System Access feature allows you to customize meter access to meet your specific security and data reporting needs.

Disclaimer to internal research use restriction:
Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Specifications
AC AdapterUniversal 50-60 Hz, 100-240 VAC power adapter (Cat. No. STARA-PWR)
Accuracy (Barometer)±6 mmHg
Accuracy (Conductivity)0.5% of reading ±1 digit > 3 μS; 0.5% of reading ±0.01 μS ≤ 3 μS
Accuracy (ISE)±0.2 mV or ±0.05% of reading whichever is greater
Accuracy (Polarographic DO)±0.2 mg/L or ±2% of reading, whichever is greater
Accuracy (RDO)±0.1 mg/L up to 8 mg/L, ±0.2 mg/L from 8 to 20 mg/L, ±10% of reading up to 50 mg/L
Accuracy (Resistivity)0.5% of reading ±1 digit
Accuracy (Salinity)0.5% of reading ±1 digit
Accuracy (TDS)0.5% of reading ±1 digit
Accuracy (mV)±0.2 mV or ±0.05% of reading whichever is greater
Accuracy (pH)±0.002
Advanced ISE FunctionsTimed endpoint, linear point to point, non-linear selectable auto-blank, low concentration range sta
Alarm OutputSet point alarm, high/low limit alarm, calibration due alarm
Backlight OptionYes
Barometric Pressure Correction400 to 850mmHg, automatic using built-in barometer or manual entry
CalibrationpH with calibration editing option, ion concentration (ISE) with calibration editing option, relativ
Calibration ModesWater-saturated air, air-saturated water, manual (Winkler titration), zero point
Calibration Points1 point relative mV (RmV) mode, 1 point ORP mode
Calibration Points (Conductivity)Up to 6
Calibration Points (ISE)1 to 6
Calibration Points (pH)1 to 6
Cell Constant0.001 to 199.9cm-1
Certifications/ComplianceCE, TUV 3-1, FCC Class A
Channels4 (Customizable with modules)
Data Management DetailsTransfer single, range or all data points to printer or computer
Data Points2000 with Date and Time Stamp
DescriptionOrion Versa Star Pro Benchtop Meter with VSTAR-ISE pH/mV/ISE/Temperature Module; VSTAR-CND Conductivity/TDS/Resistivity/Salinity/Temperature Module; VSTAR-RD RDO/DO/Temperature Module Kit
Dimensions (L x W x H)28 x 24 x 10cm (11 x 9.5 x 3.9 in.), modules add ≤ 2cm (0.8 in.) L & H
IncludesVersa Star Pro meter, VSTAR-ISE pH/ISE module, VSTAR-CND conductivity module, VSTAR-RD RDO/DO module, 8157BNUMD ROSS Ultra Triode epoxy-body pH/ATC electrode, 013005MD Orion DuraProbe 4-cell (K=0.475) conductivity sensor, 083005MD Orion polarographic dissolved oxygen sensor, 096019 Orion Star stirrer probe, 810199 ROSS pH buffer and solution kit, 011007 Orion 1413μS conductivity standard, 080513 DO maintenance kit, BOD adapter/funnel/stirrer, electrode stand, universal power adapter, literature CD, printed quick start guide, computer interface cable and meter test certificate
InputsBNC (pH or ISE), 8-pin Mini-DIN (temperature), reference pin-tip, 8-pin Mini-DIN (conductivity with temperature probe), 9-pin Mini-DIN (RDO or polarographic DO with temperature probe), 2 stirrer inputs
Isopotential Point7.000pH (adjustable), 1.000 ISE (adjustable)
KeypadNumeric with menu-specific function keys and dual purpose scroll/shortcut keys
LanguagesEnglish, Spanish, French, Italian, German, Chinese, Portuguese, Korean
Log Function TypeAutomatic data logging with Auto-Read, Timed and Single Shot measure modes; manual data logging with
MemoryNon-volatile memory preserves data log, calibration log and meter settings
Methods10 per channel with password protection
ModelOrion Versa Star Pro 90 pH/ISE/mV/RmV/ORP/Conductivity/TDS/Salinity/Resistivity/RDO Optical/Dissolved Oxygen/Temperature Benchtop Meter
OutputsUSB (2), RS232
Percent Saturation Range (Polarographic DO)0.0 to 600.0% saturation
Percent Saturation Relative Accuracy (RDO)±2% saturation ≤ 200% saturation, ±10% saturation > 200% saturation
Percent Saturation Resolution (Polarographic DO)0.1, 1% saturation
Percent Saturation Range (RDO)0.0 to 500.0% saturation
Percent Saturation Resolution (RDO)0.1, 1% saturation
Percent Saturation Relative Accuracy (Polarographic DO)±2 % saturation or ±2% of reading whichever is greater
Power RatingDC input: 9VDC, 1A
Power SupplyUniversal AC adapter (included)
Pressure Range400 to 850mmHg
Pressure Resolution0.1mmHg
Product TypeMeter; pH/Ion, Conductivity and DO; Standard Kit
Range (ISE)0.0001 to 19990 with selectable units of ppm, Molar, mg/L, %, ppb or none
Range (ORP)±2000.0 mV
Range (Polarographic DO)0.00 to 90.00mg/L
Range (RDO)0.00 to 50.00mg/L
Range (Relative mV)±2000.0 mV
Relative Humidity Range5 to 85%, non-condensing
Resolution (Conductivity)0.001 μS minimum, auto ranging up to 4 significant digits
Resolution (ISE)0.0001 minimum, 1 to 4 significant digits (user selectable)
Resolution (Polarographic DO)0.01, 0.1mg/L
Resolution (RDO)0.01, 0.1mg/L
Resolution (Resistivity)1 ohm or 0.1 meg-ohm, auto ranging
Resolution (Salinity)0.01 psu or 0.01 ppt minimum, auto ranging
Resolution (TDS)0.001 ppm minimum, auto ranging up to 4 significant digits
Resolution (mV)0.1mV
Resolution (pH)0.1/0.01/0.001
Salinity Factor0.0 to 45.0 ppt, automatic correction using conductivity reading or manual entry of sample salinity
Salinity Measurement ModesPractical salinity (psu) or natural sea water (ppt)
StabilitySelectable fast, medium, slow or Smart Stability setting; Selectable off or Automatic Smart averaging setting
TDS Factor RangeLinear (0.02 to 9.99)
Temperature Accuracy±0.1°C
Temperature CompensationLinear, nLFn (non-linear ultra pure water), nLFu (non-linear ultra pure degassed water), EP (off with warning) or off
Temperature (English) Operating41°F to 113°F (ambient)
Temperature (Metric) Operating5°C to 45°C (ambient)
Temperature (Metric) Reference5°, 10°, 15°, 20°, 25°C
Temperature Probe Calibration1 point temperature offset calibration
Temperature Range (English)22.0°F to 221.0° F
Temperature Resolution0.1°C, 0.1°F
Temperature SelectionManual or automatic with ATC temperature probe
Warranty3 year meter and module warranty
Weight (English)2.5 lb., modules add 0.5 lb. each
Weight (Metric)1.2 kg, modules add 0.2 kg each
Display TypeColor Graphic LCD
IP RatingIP54
Measurement ModespH, mV, Relative mV, ORP, Ion Concentration (ISE) with Temperature, TDS, Conductivity, Salinity
Range (Conductivity)0.001 μS/cm to 3000 mS/cm
Range (Resistivity)2 ohm to 100.0 meg-ohm
Range (Salinity)0.06 to 80.00 psu, 0.05 to 42.00 ppt
Range (TDS)0.001 ppm to 200.0 ppt
Range (mV)±2000.0 mV
Range (pH)-2 to 20
Temperature Range (Metric)-5.0°C to +105.0°C
Unit SizeEach