WesternDot® 625 Goat-Anti Chicken Secondary Antibody (VIVID® Technology)

Catalog number: W10827

Invitrogen™

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

WesternDot® antibodies deliver superior signal-to-noise ratios and exhibit comparable sensitivity to ECL-based chemiluminescent methods in western blot applications. No stripping and re-probing or multiple blots are required for multiple protein detection as commonly practiced with ECL or colorimetric detection. Different colors of WesternDot® antibodies can be applied to a single blot and multiple proteins detected simultaneously using standard gel or blot imaging platforms. Our WesternDot® antibodies use Qdot® nanocrystals enhanced with VIVID® technology, which are brighter than the original Qdot® reagents. Additionally, they have reduced intensity differences between the different colors, simplifying multiplex applications. Specific information about compatible imagers can be found in the product manual below.

Easy to detect on ethidium bromide gel doc imaging systems
Reduce time to result—eliminate cumbersome HRP and ECLl optimization
Easy to switch—WesternDot® antibodies use standard membranes, blocking solutions, and buffers
WesternDot® 625 antibody optimal excitation/emission: 405-485/625 nm

Learn more about WesternDot® antibodies
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Reactivity: Chicken
Target Isotype: IgG
Host Species: Goat
Label or Dye: Qdot® 625
Product Line: VIVID®, WesternDot®
Product Size: 25 blots
Shipping Condition: Room Temperature

Contents & storage

Store in refrigerator (2–8°C). Do not freeze.

Documents

Manuals & protocols

Recommended products