This product ships via ground from US northeast.
Water, UHPLC-MS
Water, UHPLC-MS
Water, UHPLC-MS
Water, UHPLC-MS
Water, UHPLC-MS
Thermo Scientific™

Water, UHPLC-MS

Unique signal-to-noise specification for the chemical industry where the solvent quality is linked directly to the sensitivity of the mass spectrometer. | CAS: 7732-18-5 | H2O | 18.015 g/mol
Have Questions?
Catalog NumberQuantityPackaging
W811 LClear Borosilicate Glass Bottle
Catalog number W81
Price (USD)
43.65
Special offer
Online exclusive
Ends: 28-Dec-2026
51.75
Save 8.10
Each
Quantity:
1 L
Packaging:
Clear Borosilicate Glass Bottle
Price (USD)
43.65
Special offer
Online exclusive
Ends: 28-Dec-2026
51.75
Save 8.10
Each

Water, H2O, CAS Number-7732-18-5, dihydrogen oxide, dihydrogen monoxide, 1L, <-0.01 AU at 230nm, <-0.01 AU at 210nm, <-0.01 AU at 220nm, <-0.01 AU at 240nm, <-0.005 AU at 280nm, <-0.005 AU at 260nm, <-0.005 AU at 254nm, 2ppb max., 100 deg.C, 5ppb max., CHEBI:15377, Colorless, 1.000g/cm3, UHPLC-MS

RUO – Research Use Only
Specifications
Absorbance≤0.005 AU at 254nm, ≤0.005 AU at 260nm, ≤0.005 AU at 280nm, ≤0.01 AU at 210nm, ≤0.01 AU at 220nm, ≤0.01 AU at 230nm, ≤0.01 AU at 240nm
Boiling Point100°C
CAS7732-18-5
ChEBICHEBI:15377
Chemical Name or MaterialWater
ColorColorless
Density1.000 g/cm3
IUPAC Nameoxidane
InChI KeyXLYOFNOQVPJJNP-UHFFFAOYSA-N
Melting Point0°C
Molecular FormulaH2O
PackagingClear Borosilicate Glass Bottle
Physical FormLiquid
PubChem CID962
Recommended StorageRoom temperature
Residue after Evaporation≤1ppm
SMILESO
Synonymdihydrogen oxide, dihydrogen monoxide
Vapor Pressure17.5mmHg at 20°C
GradeUHPLC-MS
Molecular Weight (g/mol)18.015
Quantity1 L
pH7
Unit SizeEach