NuPAGE™ Bis-Tris Midi Protein Gels, 4 to 12%, 1.0 mm, WedgeWell™ format
NuPAGE™ Bis-Tris Midi Protein Gels, 4 to 12%, 1.0 mm, WedgeWell™ format
NuPAGE™ Bis-Tris Midi Protein Gels, 4 to 12%, 1.0 mm, WedgeWell™ format
Invitrogen™

NuPAGE™ Bis-Tris Midi Protein Gels, 4 to 12%, 1.0 mm, WedgeWell™ format

Invitrogen NuPAGE Bis-Tris protein gels, WedgeWell format are precast polyacrylamide gels that provide broad molecular weight protein separation with high resolution and sample integrity. These precast gels are ideal for applications where protein integrity is crucial.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호
WBT41226BOX26-well
WBT41212BOX12 + 2-well
WBT41220BOX20-well
카탈로그 번호 WBT41226BOX
제품 가격(KRW)
321,000
온라인 행사
Ends: 30-Sep-2026
377,000
할인액 56,000 (15%)
10 gels (1 box)
웰:
26-well
제품 가격(KRW)
321,000
온라인 행사
Ends: 30-Sep-2026
377,000
할인액 56,000 (15%)
10 gels (1 box)
Invitrogen NuPAGE Bis-Tris protein gels, WedgeWell format are precast polyacrylamide gels that provide broad molecular weight protein separation with high resolution and sample integrity. These precast gels are ideal for applications where protein integrity is crucial. Unlike traditional Tris-glycine gels, NuPAGE Bis-Tris gels have a neutral pH environment that minimizes protein modifications. Use NuPAGE Bis-Tris gels for preparing proteins for sequencing, mass spectrometry, and any other techniques where protein integrity is important.

Invitrogen NuPAGE Bis-Tris WedgeWell format protein gels are precast polyacrylamide gels that provide broad molecular weight protein separation with high resolution and sample integrity. These precast gels are ideal for applications where protein integrity is crucial. Unlike traditional Tris-glycine gels, NuPAGE Bis-Tris gels have a neutral pH environment that minimizes protein modifications. Use NuPAGE Bis-Tris gels for preparing proteins for sequencing, mass spectrometry, and any other techniques where protein integrity is important.

Product features

  • Better protein integrity—optimized sample preparation process preserves your proteins
  • Faster run times—get separation of your proteins in as little as 30 minutes
  • Longer shelf life—NuPAGE Bis-Tris gels can be stored for 12 months at room temperature
  • Wedge-shaped wells—for easier sample loading and up to two times higher sample volume

Choose the right NuPAGE Bis-Tris gel for your protein separation

NuPAGE Bis-Tris gels are formulated for denaturing gel electrophoresis applications. For optimal sample preparation, use the NuPAGE LDS Sample Buffer (NP0007) and NuPAGE Sample Reducing Agent (NP0004). NuPAGE Antioxidant (NP0005) can be added to the running buffer to maintain the reduced state of the proteins during the run allowing for maximum band sharpness if desired. The gels can be run using NuPAGE MES SDS Running Buffer (NP0002) to better resolve small proteins with a faster run time or NuPAGE MOPS SDS Running Buffer (NP000102) to resolve medium- to large-size proteins.

NuPAGE WedgeWell Bis-Tris Midi gels can be run in the SureLock Tandem Midi Gel Tank (STM1001) or XCell4 SureLock Midi-Cell (WR0100).

For transfer of proteins to a membrane, rapid semi-dry transfer can be done using the Invitrogen Power Blotter or rapid dry transfer using the iBlot 3 Gel Transfer Device (IB31001). Alternatively, traditional wet transfer can be performed using the SureLock Tandem Midi Blot Module (STM2001) using NuPAGE Transfer buffer (NP00061).

Convenient to use

NuPAGE Bis-Tris WedgeWell format gels have innovative wedge-shaped wells that allow up to two times more sample load volume than other 1.0 mm-thick gels. The wedge-shaped wells also have larger openings, making sample loading easier.

For Research Use Only. Not for use in diagnostic procedures.
사양
용도(장비)SureLock Tandem Midi Gel Tank
Gel Thickness1.0 mm
길이(미터법)8 cm
분리 모드Molecular Weight
수량10 gels (1 box)
샘플 로딩 부피Up to 40 μL
유통 기한12 Months
배송 조건Room Temperature
폭(미터법)13 cm
젤 비율4 to 12%
젤 크기Midi
젤 유형Bis-Tris
제품라인NuPAGE
분리 범위3.5 to 260 kDa
분리 유형Denaturing
26-well
Unit Size10 gels (1 box)
구성 및 보관
Store at 4–25°C. Do not freeze.

자주 묻는 질문(FAQ)

Can I prepare my protein sample with the reducing agent and store it for future use?

DTT is not stable, so it must be added and the reduction performed just prior to loading your samples.

Find additional tips, troubleshooting help, and resources within our Protein Gel 1D Electrophoresis Support Center.

My LDS or SDS sample buffer precipitates when stored at 4 degrees C. Can I warm it up? Can I store it at room temperature?

Precipitation of the LDS or SDS at 4 degrees C is normal. Bring the buffer to room temperature and mix until the LDS/SDS goes into solution. If you do not want to wait for it to dissolve, you can store the sample buffer at room temperature.

Find additional tips, troubleshooting help, and resources within our Protein Gel 1D Electrophoresis Support Center.

How are Bolt gels different than NuPAGE gels?

While they are both Bis-Tris based gels, the chemistries are very different since Bolt gels are optimized for western blotting. Another key difference is the wedge well design of the Bolt gels, which allows larger sample volumes to be loaded.

Find additional tips, troubleshooting help, and resources within our Protein Gel 1D Electrophoresis Support Center.

Do I need to add water or buffer to the Displacement Dam when I run one or two gels in the XCell4 SureLock Midi-Cell?

The XCell4 Displacement Dam is used when fewer than 3 gels are run in the XCell4 SureLock Midi-Cell. The Displacement Dam is placed in the lower buffer chamber instead of a second buffer core. Do not add any buffer or water to the Displacement Dam. However, if water or buffer is accidentally added to the Displacement Dam, it does not affect the electrophoresis run.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

What are the maximum load volumes for the NuPAGE Midi Gels?

Recommended maximum load volumes:
12 + 2-well gel: 45 µL in sample well, 15 µL in marker well
20-well gel: 25 µL
26-well gel: 15 µL

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.