Empty Gel Cassettes with Combs, Midi, 26-well
Invitrogen™
Empty Gel Cassettes with Combs, Midi, 26-well
These empty gel cassettes are pre-assembled, single-use, empty plastic cassettes with combs that allow you to pour your own midi-sizeRead more
Have Questions?
Catalog NumberQuantity
WC10261 pack
Catalog number WC1026
Price (USD)
106.65
Online exclusive
112.00
Save 5.35
1 pack
Quantity:
1 pack
Price (USD)
106.65
Online exclusive
112.00
Save 5.35
1 pack
These empty gel cassettes are pre-assembled, single-use, empty plastic cassettes with combs that allow you to pour your own midi-size gels. An XCell4 SureLock™ Midi-Cell Runner is required to run the gels. The cassettes are sealed on three sides to prevent leaking and do not require the use of a special casting stand. Gels poured in these cassettes are 1.0 mm thick. The cassettes have printed well outlines for easier gel loading.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)XCell4 SureLock Midi-Cell
Gel Thickness1.0 mm
Length (Metric) Gel8 cm
Quantity1 pack
Running DimensionVertical
Shipping ConditionRoom Temperature
Width (Metric) Gel13 cm
Gel SizeMidi
Product TypeEmpty Gel Cassette
Wells26-well
Unit Size1 pack
Contents & Storage
10 cassettes (with 26-well combs) per pack
Frequently asked questions (FAQs)
Do you offer empty cassettes and combs for casting Mini and Midi protein gels?
We do offer disposable, empty plastic cassettes and combs to provide the option of pouring your own Mini or Midi gels for use with the XCell SureLock Mini-Cell, Mini Gel Tank, or XCell4 SureLock Midi-Cell, respectively. All cassettes are sealed on three sides to prevent leaking. Since the cassettes are pre-sealed with the slot pre-taped, a bulky casting stand is not necessary. The empty cassettes and combs can be found here (https://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-gel-electrophoresis/power-supplies-protein-electrophoresis.html#staining).
- The Empty Mini Gel Cassettes are available in two thicknesses (1.0 mm and 1.5 mm) and can be used with Invitrogen Combs that are available in two thicknesses (1.0 mm and 1.5 mm), and in 7 types (1 well, 2 well, 5 well, 10 well, 12 well, 15 well, and 2D well).
- The Bolt Empty Mini Gel Cassettes are available in 1.0 mm thickness and can be used with Bolt 10- or 12-well combs.
- The Empty Midi Gel Cassettes are available in 1.0 mm thickness and can be used with three Invitrogen Comb types (12 + 2 wells, 20 wells, and 26 wells).
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.