Gibco™

Rat (Wistar) Cryopreserved Hepatocytes, Plateable Female

Catalog number:  WICP20

Related applications:

Pharma & Biopharma

| Primary Cell Culture

Back to top

Description

These plateable cryopreserved female rat (Wistar) hepatocytes have been characterized for general Phase I and Phase II drug metabolizing enzyme activities.

Cells per vial: 4–8 million

Get the Results You Need with GIBCO® Hepatocytes
High quality hepatocytes with high viabilities, in vivo-like enzymee xpression levels, and proper cell morphology increase the ability to draw in vitroin vivo correlations and make sound decisions regarding a compound's fate. Experienced technicians and proprietary isolation techniques combined with stringent release specifications ensure that you receive the highest quality cryopreserved hepatocytes.

Each lot is tested for:

• Phase I and II enzyme activities
• Proper morphology
• Viabilities generally >75%
• Attachment efficiency
• Monolayer formation and integrity

For research use only. Not intended for human or animal therapeutic or diagnostic use.

Related Links:

Learn more about our cryopreserved hepatocytes.

Learn about our full line of hepatocytes.

Note:
These products require shipment in LN2 vapor dewars therefore additional fees may apply.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Cell Type: Hepatocytes
Donor Source: Pooled
Form: Cryopreserved
Product Line: Gibco®
Product Size: 1 mL
Species: Rat

Contents & storage

Cryopreserved Hepatocytes stored in liquid nitrogen (LN2).