Secondary Antibody Solution Alk-Phos. Conjugated (Anti-Rabbit)
Invitrogen™

Secondary Antibody Solution Alk-Phos. Conjugated (Anti-Rabbit)

The Secondary Alkaline-Phoshatase conjugated (Anti-Rabit) Antibody solution is a pre-diluted, ready-to-use formulation containing alkaline phosphatase labeled polyclonal antibodies in aRead more
Have Questions?
Catalog NumberQuantity
WP20007100 mL (2 Bottles)
Catalog number WP20007
Price (USD)
140.60
Special offer
Online exclusive
Ends: 27-Jun-2026
148.00
Save 7.40 (5%)
2 bottles
Quantity:
100 mL (2 Bottles)
Price (USD)
140.60
Special offer
Online exclusive
Ends: 27-Jun-2026
148.00
Save 7.40 (5%)
2 bottles
The Secondary Alkaline-Phoshatase conjugated (Anti-Rabit) Antibody solution is a pre-diluted, ready-to-use formulation containing alkaline phosphatase labeled polyclonal antibodies in a proprietary stabilizing reagent. A goat anti-rabbit polyclonal alk-phos conjugate, this Secondary antibody is compatable with both chemiluminescent and chromogenic methods of immunodetection, and will identify antigens bound with a primary rabbit antibody.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Primary or SecondarySecondary
ReactivityRabbit
Shipping ConditionWet Ice
Whole MoleculeWhole Antibody
ClassificationPolyclonal
ConjugateAlkaline Phosphatase (AP)
FormLiquid
Host SpeciesGoat
IsotypeIgG
Quantity100 mL (2 Bottles)
Target SpeciesRabbit
Unit Size2 bottles
Contents & Storage
Store in refrigerator (2–8°C).

Frequently asked questions (FAQs)

Can I purchase the Secondary Antibody solutions from the WesternBreeze Chemiluminescent Detection kits as standalone products?

The goat anti-mouse and goat anti-rabbit secondary antibody solutions AP-conjugated are available as standalone products (Cat. Nos. WP20006 and WP20007), but the rabbit anti-goat secondary antibody solution AP-conjugated is not available as a standalone product.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Can I purchase the Secondary Antibody solutions from the WesternBreeze Chromogenic Detection kits as standalone products?

The goat anti-mouse and goat anti-rabbit secondary antibody solutions AP-conjugated are available as standalone products (Cat. Nos. WP20006 and WP20007) but the rabbit anti-goat secondary antibody solution AP-conjugated is not available as a standalone product.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.