SURELOCK XCELL 4 DUMMY - FAQs
SURELOCK XCELL 4 DUMMY - FAQs
View additional product information for SURELOCK XCELL 4 DUMMY - FAQs (WR0106)
3 product FAQs found
When should I use the XCell4 Displacement Dam with the XCell4 SureLock Midi-Cell?
Use the XCell4 Displacement Dam when you wish to run less than 3 gels in the XCell4 SureLock Midi-Cell. The Displacement Dam is placed in the Lower Buffer Chamber instead of a second Buffer Core. Do not add any buffer or water to the Displacement Dam. However, if water or buffer is accidentally added to the Displacement Dam, it does not affect the electrophoresis run.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
Can I purchase replacement parts for the XCell4 SureLock Midi-Cell?
Here are the replacement parts we offer for the XCell4 SureLock Midi-Cell. They can only be ordered by phone:
- XCell4 SureLock Midi-Cell Lid: Cat. No. WR0101
- XCell4 SureLock Midi-Cell Lower Buffer Chamber: Cat. No. WR0102
- XCell4 SureLock Midi-Cell Displacement Dam: Cat. No. WR0106
- XCell SureLock Upgrade kit: Cat. No. WR0112 (contains 1 SureLock Assembly, 1 buffer dam, and 2 cores) Please note that in addition to ordering on the phone by calling Customer Service, this item can also be ordered using the “Quick Order” link (https://www.thermofisher.com/store/quick-order) that can be found at the top right of most pages on our website. The price will display when the item is added to your shopping cart.
Find additional tips, troubleshooting help, and resources within our Protein Gel Electrophoresis Chambers, Power Supplies, and Accessories Support Center.
What are the components of the XCell4 SureLock Midi-Cell?
The XCell4 SureLock Midi-Cell includes the lower buffer chamber, lid with cables, two buffer cores, buffer dam, displacement dam, SureLock assembly, and gel knife. For descriptions and images of the various parts, please refer to pages 1-3 of the product manual (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/xcell4_midicell_man.pdf).
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.