NuPAGE™ Tris-Acetate Midi Protein Gels, 3 to 8%, 1.0 mm, WedgeWell™ format, 26-well, 10 gels (1 box) - FAQs
NuPAGE™ Tris-Acetate Midi Protein Gels, 3 to 8%, 1.0 mm, WedgeWell™ format, 26-well, 10 gels (1 box) - FAQs
View additional product information for NuPAGE™ Tris-Acetate Midi Protein Gels, 3 to 8%, 1.0 mm, WedgeWell™ format - FAQs (WTA03820BOX, WTA03826BOX, WTA03812BOX)
1 product FAQs found
What are the recommended maximum sample loading volumes for the new WedgeWell format holds for the midi gels?
For the 12+2 well large gels, the maximum sample loading volume is 100 µL. For the 12+2 well small gels, the maximum sample loading volume is 90 µL. For 20 well gels, the maximum sample loading volume is 60 µL and for the 26 well gels, the maximum sample loading volume is 40 µL.
Find additional tips, troubleshooting help, and resources within our Protein Gel 1D Electrophoresis Support Center.