Novex™ 6% Tris-Glycine Mini Gels, WedgeWell™ format, 10-well - FAQs
Novex™ 6% Tris-Glycine Mini Gels, WedgeWell™ format, 10-well - FAQs
View additional product information for Novex™ 6% Tris-Glycine Mini Gels, WedgeWell™ format, 10-well - FAQs (XP00060BOXPR)
This product does not have any FAQs available.