Novex™ 10% Tris-Glycine Mini Gels, WedgeWell™ format, 12-well - FAQs
Novex™ 10% Tris-Glycine Mini Gels, WedgeWell™ format, 12-well - FAQs
View additional product information for Novex™ 10% Tris-Glycine Mini Gels, WedgeWell™ format, 12-well - FAQs (XP00102BOXPR)
This product does not have any FAQs available.