Novex™ 10% Tris-Glycine Mini Gels, WedgeWell™ format, 15-well - FAQs
Novex™ 10% Tris-Glycine Mini Gels, WedgeWell™ format, 15-well - FAQs
View additional product information for Novex™ 10% Tris-Glycine Mini Gels, WedgeWell™ format, 15-well - FAQs (XP00105BOXPR)
This product does not have any FAQs available.