Novex™ 4-12% Tris-Glycine Mini Gels, WedgeWell™ format, 10-well - FAQs
Novex™ 4-12% Tris-Glycine Mini Gels, WedgeWell™ format, 10-well - FAQs
View additional product information for Novex™ 4-12% Tris-Glycine Mini Gels, WedgeWell™ format, 10-well - FAQs (XP04120BOXPR)
4 product FAQs found
Is it okay to use protein gels past their expiration date?
We do not recommend using gels past their expiration date because over time, the polyacrylamide hydrolyzes to acrylic acid and ammonia and this will affect the resolution of the proteins. Breakdown of polyacrylamide matrix is identified by:
- Ghost bands and doublets, seen first in the high molecular weight proteins
- Smiling of dye front across the gel, with bands in outer lanes becoming very slanted - proteins run slower there due to change in pH and pore size over time.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
Can I run your protein gels overnight?
This is not really recommended, but it is always possible to increase run time by lowering the voltage of the run. In general, the relationships are linear - i.e., decreasing voltage by half will double the run time.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
Do I need to increase the voltage when I run a 1.5 mm protein gel versus a 1.0 mm gel?
If you are running the gel at constant voltage, you do not need to increase the voltage regardless of the thickness of the gel.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
Why do you recommend running your protein gels at constant voltage?
Using constant voltage allows the current and power to decrease during the run, providing a safety margin in case of a break in the system. Having lower power is a safety benefit and will also decrease the chances of experiencing an overheating of the gel. Further, the constant voltage setting does not need adjustment to account for differences in number or thickness of gels being run.
Find additional tips, troubleshooting help, and resources within ourProtein Gel 1D Electrophoresis Support Center.