Attention: Novex™ Value™ 4 to 12%, Tris-Glycine, 1.0 mm, Mini Protein Gel, 15-well (Catalog number: XV04125PK20) is planned for discontinuation. The alternate product is Catalog number: XP04125BOX. Please contact Technical Support with any questions.
Novex™ Value™ Tris-Glycine Mini Protein Gel, 4 to 12%, 1.0 mm, 15-well
Invitrogen™

Novex™ Value™ Tris-Glycine Mini Protein Gel, 4 to 12%, 1.0 mm, 15-well

Novex™ Value Tris-Glycine Mini Gels provide a great balance of price and performance for your protein separation needs. Novex ValueRead more
Have Questions?
Catalog NumberQuantity
XV04125PK2020 gels (1 box)
Catalog number XV04125PK20
Price (USD)
217.65
Online exclusive
229.00
Save 11.35
20 gels (1 box)
Quantity:
20 gels (1 box)
Price (USD)
217.65
Online exclusive
229.00
Save 11.35
20 gels (1 box)
Novex™ Value Tris-Glycine Mini Gels provide a great balance of price and performance for your protein separation needs. Novex Value Tris-Glycine polyacrylamide gels provide good separation and resolution of your proteins in an affordable gel that is based on traditional Laemmli protein electrophoresis. Each box contains 20 gels.

Features of the Novex Value gels include:
• Excellent balance of value and performance—great for qualitative analysis (e.g., screening, protein purification verification)
• Fast run conditions—separate your proteins using constant voltage in less than 60 minutes
• Options for native and denaturing protein assays

Choose the right gel format for your experiments
Novex Value Tris-Glycine gels come in a fixed 10% option as well 4–12% and 4–20% gradients.

Run your proteins in native or denatured form
Novex Value Tris-Glycine gels do not contain SDS and can be used to run your proteins in native or denatured form. For denatured proteins, we recommend using Novex Tris-Glycine SDS Sample Buffer and Novex Tris-Glycine SDS Running Buffer. For native proteins, we recommend using Novex Tris-Glycine Native Sample Buffer and Novex Tris-Glycine Native Running Buffer.

For transferring your proteins to membrane, we recommend using Novex Tris-Glycine Transfer Buffer. Transfers can be done using the Mini Blot Module or the iBlot™ Gel Transfer Device.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Description4-12% Value Tris-Glycine Gels, 1.0 mm, 15-well, Box of 20
For Use With (Application)Protein Gel Electrophoresis
For Use With (Equipment)XCell SureLock Mini-Cell
Gel Thickness1.0 mm
Length (Metric)8 cm
Length (Metric) Gel8 cm
MaterialTris-Glycine Gel
Mode of SeparationMolecular Weight
Quantity20 gels (1 box)
Recommended ApplicationsDenaturing, Native
Sample Loading Volumeup to 15 μL
Shelf Life12 Months
Shipping ConditionApproved for shipment at Room Temperature or on Wet Ice
Storage RequirementsStore at 4°C. Shelf life is 12 months.
Thickness1.0 mm
Well Design1D Well
Width (Metric)8 cm
Width (Metric) Gel8 cm
Gel Percentage4 to 12%
Gel SizeMini
Gel TypeTris-Glycine
Product LineNovex Value
Product TypeGradient Page
Separation Range20 to 250 kDa
Separation TypeDenaturing, Native
Separation ofProtein
Wells15-well
Unit Size20 gels (1 box)
Contents & Storage
Store in refrigerator (2–8°C).
Shelf life up to 12 months depending on gel type.

Frequently asked questions (FAQs)

Can Novex WedgeWell gels be stored at room temperature?

No. Novex WedgeWell gels must be stored at 4 degrees C.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

Do the Novex WedgeWell gels require any specific buffers?

The Novex WedgeWell gels can use standard Tris-Glycine sample and running buffers. For running under native conditions, we recommend using sample and running buffers that do not contain SDS (such as our Native Tris-Glycine premade buffers).

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

With Novex WedgeWell gels, what in the chemistry changed to allow for a longer shelf life?

We were able to optimize shelf life through a proprietary gel formulation change. Unfortunately, we are unable to provide specific details on the chemical changes.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

What is the orientation of the printing on the cassette for the Novex WedgeWell gels?

We use what we call ‘mirror printing', the same way we do for Bolt gels. This means that when the gel is loaded in the Mini Gel Tank, the text will face in the readable direction (left to right).

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

Can the new Novex WedgeWell gels be run in the XCell SureLock Mini-Cell?

Yes, the new Novex WedgeWell gels can be run in the XCell SureLock Mini-Cell.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.