Attention: Novex™ Value™ 4 to 20%, Tris-Glycine, 1.0 mm, Mini Protein Gel, 10-well (Catalog number: XV04200PK20) is planned for discontinuation. The alternate product is Catalog number: XP04200BOX. Please contact Technical Support with any questions.
Novex™ Value™ Tris-Glycine Mini Protein Gel, 4 to 20%, 1.0 mm, 10-well
Invitrogen™

Novex™ Value™ Tris-Glycine Mini Protein Gel, 4 to 20%, 1.0 mm, 10-well

Novex™ Value Tris-Glycine Mini Gels provide a great balance of price and performance for your protein separation needs. Novex ValueRead more
Have Questions?
Catalog NumberQuantity
XV04200PK2020 gels (1 box)
Catalog number XV04200PK20
Price (USD)
224.65
Online exclusive
229.00
Save 4.35
20 gels (1 box)
Quantity:
20 gels (1 box)
Price (USD)
224.65
Online exclusive
229.00
Save 4.35
20 gels (1 box)
Novex™ Value Tris-Glycine Mini Gels provide a great balance of price and performance for your protein separation needs. Novex Value Tris-Glycine polyacrylamide gels provide good separation and resolution of your proteins in an affordable gel that is based on traditional Laemmli protein electrophoresis. Each box contains 20 gels.

Features of the Novex Value gels include:
• Excellent balance of value and performance—great for qualitative analysis (e.g., screening, protein purification verification)
• Fast run conditions—separate your proteins using constant voltage in less than 60 minutes
• Options for native and denaturing protein assays

Choose the right gel format for your experiments
Novex Value Tris-Glycine gels come in a fixed 10% option as well 4–12% and 4–20% gradients.

Run your proteins in native or denatured form
Novex Value Tris-Glycine gels do not contain SDS and can be used to run your proteins in native or denatured form. For denatured proteins, we recommend using Novex Tris-Glycine SDS Sample Buffer and Novex Tris-Glycine SDS Running Buffer. For native proteins, we recommend using Novex Tris-Glycine Native Sample Buffer and Novex Tris-Glycine Native Running Buffer.

For transferring your proteins to membrane, we recommend using Novex Tris-Glycine Transfer Buffer. Transfers can be done using the Mini Blot Module or the iBlot™ Gel Transfer Device.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionNovex™ Value™ 4 to 20% Tris-Glycine Mini Gels, 20 Gels, 0.039 in. Thickness, 8 x 8cm Gel Size, Gel, For Protein Gel Electrophoresis, 12 Months Shelf Life, 2 to 8°C Storage Temperature
For Use With (Application)Protein Gel Electrophoresis
For Use With (Equipment)Mini Gel Tank
FormulationGel
Gel Thickness1.0 mm
Length (Metric)8 cm
Length (Metric) Gel8 cm
MaterialTris-Glycine
Mode of SeparationMolecular Weight
Quantity20 gels (1 box)
Recommended ApplicationsDenaturing, Native
Sample Loading Volumeup to 25 μL
Shelf Life12 Months
Shipping ConditionApproved for shipment at Room Temperature or on Wet Ice
Storage RequirementsStore at 4°C. Shelf life is 12 months.
Thickness1.0 mm
Thickness (English)0.039 in.
Thickness (Metric)1.0 mm
Well Design1D Well
Width (Metric)8 cm
Width (Metric) Gel8 cm
Gel Percentage4 to 20%
Gel SizeMini
Gel TypeTris-Glycine
Product LineNovex Value
Product TypeGradient Page
Separation Range20 to 200 kDa
Separation TypeDenaturing, Native
Separation ofProtein
Wells10-well
Unit Size20 gels (1 box)
Contents & Storage
Store in refrigerator (2–8°C).
Shelf life up to 12 months depending on gel type.

Frequently asked questions (FAQs)

I am transferring a Tris-Glycine gel using constant voltage and the current reading is way over the expected starting current. Can you offer some suggestions?

The most common cause of abnormally high current is the transfer buffer. If the transfer buffer is too concentrated, this leads to increased conductivity and current. High current may also occur if Tris-HCl is accidentally substituted for the Tris base required in the transfer buffer. This will again result in low buffer pH and lead to increased conductivity and current and subsequently, overheating. We recommend checking the transfer buffer and its reagent components and re-diluting or remaking the buffer.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.