Novex™ Value™ Tris-Glycine Mini Protein Gel, 4 to 20%, 1.0 mm, 15-well - FAQs
Novex™ Value™ Tris-Glycine Mini Protein Gel, 4 to 20%, 1.0 mm, 15-well - FAQs
View additional product information for Novex™ Value™ Tris-Glycine Mini Protein Gel, 4 to 20%, 1.0 mm, 15-well - FAQs (XV04205PK20)
1 product FAQs found
I am transferring a Tris-Glycine gel using constant voltage and the current reading is way over the expected starting current. Can you offer some suggestions?
The most common cause of abnormally high current is the transfer buffer. If the transfer buffer is too concentrated, this leads to increased conductivity and current. High current may also occur if Tris-HCl is accidentally substituted for the Tris base required in the transfer buffer. This will again result in low buffer pH and lead to increased conductivity and current and subsequently, overheating. We recommend checking the transfer buffer and its reagent components and re-diluting or remaking the buffer.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.