alamarBlue™ Dye
alamarBlue™ Dye
Thermo Scientific™
alamarBlue™ Dye
Allows three assays to be performed with just one dye, including cell growth determination, in vitro cytotoxicity and cytokine assays.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
Y0001010 mL
Y00-02525 mL
Y00-100100 mL
Catalog number Y00010
Price (USD)
-
Request A Quote
Quantity:
10 mL
Thermo Scientific™ alamarBlue provides unmatched sensitivity and versatility, allowing three assays to be performed with just one dye, including cell growth determination, in vitro cytotoxicity and cytokine assays.
alamarBlue, a resazurin-based product with extended incubation time, is stable for up to 5 days.
Specifications
DescriptionalamarBlue indicator dye
Quantity10 mL
Unit SizeEach
Frequently asked questions (FAQs)
How do alamarBlue reagent and PrestoBlue reagent differ from resazurin and C12-resazurin?
alamarBlue reagent and PrestoBlue reagent contain resazurin in a proprietary stabilizing formulation that allows for a convenient “mix, incubate, and read” protocol. PrestoBlue reagent is an improvement in the formulation of alamarBlue reagent that allows for much faster staining (typically 10 minutes vs. 1-4 hours to obtain a similar signal and sensitivity). C12-resazurin is a derivative of resazurin that has better cellular retention and thus allows for analysis on a flow cytometer and multiplexing with viability indicators and other biomarkers.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.