Zymosan A (S. cerevisiae) BioParticles™, Alexa Fluor™ 488 conjugate - FAQs
Zymosan A (S. cerevisiae) BioParticles™, Alexa Fluor™ 488 conjugate - FAQs
View additional product information for Zymosan A (S. cerevisiae) BioParticles™, Alexa Fluor™ 488 conjugate - FAQs (Z23373)
4 product FAQs found
Are the Invitrogen BioParticles products sterile?
While the bacteria have been attenuated with formaldehyde and alcohol desiccation, the BioParticles products are not considered sterile, and we do not recommend incubation of more than 4 hours. This applies to all of our dye-labeled (pHrodo, Alexa Fluor, etc.) and unlabeled BioParticles products.
What is the type of bond that attaches the dyes to the BioParticles probes?
We use amine-reactive dyes to covalently attach fluorescent dyes to all of our BioParticles probes such as the Escherichia coli (K-12 strain) BioParticles probes, Staphylococcus aureus (Wood strain without protein A) BioParticles, and the Zymosan A (S. cerevisiae) BioParticles probes.
What can the BioParticles product line be used for?
Fluorescent Bioparticles have been employed to study phagocytosis by fluorescence microscopy, quantitative spectrofluorometry, and flow cytometry. We offer E.Coli, S. aureus, and zymosan BioParticles conjugates covalently labeled with a variety of different fluorophores.
What is the fluorescence excitation/emission of AlexaFluor 488 in the Zymosan A Bioparticles kit?
The fluorescence excitation/emission of AlexaFluor 488 in buffer at pH 8 is 495/519 nm.