Zenon™ Allophycocyanin Mouse IgG1 Labeling Kit, 25 Reactions kit - FAQs
Zenon™ Allophycocyanin Mouse IgG1 Labeling Kit, 25 Reactions kit - FAQs
View additional product information for Zenon™ Mouse IgG1 Labeling Kits - FAQs (Z25051, Z25006, Z25005, Z25008, Z25007, Z25002, Z25013, Z25052, Z25011, Z25055)
1 product FAQs found
How should I store my allophycocyanin (APC)-conjugated antibody?
Allophycocyanin-conjugated antibodies must be stored at 2-8 degrees C. Antibodies conjugated to phycobiliproteins (APC, R-PE) must never be frozen. Once frozen, even briefly, they are no longer usable.