Zenon™ Allophycocyanin Rabbit IgG Labeling Kit, 25 Reactions kit - FAQs
Zenon™ Allophycocyanin Rabbit IgG Labeling Kit, 25 Reactions kit - FAQs
View additional product information for Zenon™ Rabbit IgG Labeling Kits - FAQs (Z25307, Z25306, Z25308, Z25303, Z25302, Z25313, Z25305, Z25355, Z25351)
1 product FAQs found
How should I store my allophycocyanin (APC)-conjugated antibody?
Allophycocyanin-conjugated antibodies must be stored at 2-8 degrees C. Antibodies conjugated to phycobiliproteins (APC, R-PE) must never be frozen. Once frozen, even briefly, they are no longer usable.