Zenon™ Allophycocyanin Human IgG Labeling Kit, 25 Reactions kit - FAQs
Zenon™ Allophycocyanin Human IgG Labeling Kit, 25 Reactions kit - FAQs
View additional product information for Zenon™ Human IgG Labeling Kits - FAQs (Z25408, Z25407, Z25402, Z25455, Z25451)
1 product FAQs found
How should I store my allophycocyanin (APC)-conjugated antibody?
Allophycocyanin-conjugated antibodies must be stored at 2-8 degrees C. Antibodies conjugated to phycobiliproteins (APC, R-PE) must never be frozen. Once frozen, even briefly, they are no longer usable.