Zymosan A <i>S. cerevisiae </i> BioParticles&trade;, unlabeled
Invitrogen™

Zymosan A S. cerevisiae BioParticles™, unlabeled

The Molecular Probes™ unlabeled BioParticles™ product line comprises heat- or chemically killed S. aureus (Wood strain, without protein A) andRead more
Have Questions?
Catalog NumberQuantity
Z2849100 mg
Catalog number Z2849
Price (USD)
146.00
100 mg
Quantity:
100 mg
Price (USD)
146.00
100 mg
The Molecular Probes™ unlabeled BioParticles™ product line comprises heat- or chemically killed S. aureus (Wood strain, without protein A) and zymosan (S. cerevisiae) cells. These nonfluorescent BioParticles™ products are useful in phagocytosis assays, both as controls and for custom-labeling with reactive dyes.

BioParticles Specifications:
• Label (Ex/Em): Unlabeled
• Particle: Zymosan (S. cerevisiae)
• Opsonizing reagent available


Using BioParticles™ Products
BioParticles™ conjugates are provided as lyophilized powders. There are approximately 3 x 108 S. aureus particles per mg solid and approximately 2 x 107 zymosan particles per mg solid. BioParticles™ conjugates can be reconstituted in the buffer of your choice for use in phagocytosis assays.

Find More BioParticles™ Products
We offer a large range of dye-labeled and unlabeled E. coli (K-12 strain), S. aureus (Wood strain, without protein A), and zymosan (S. cerevisiae) BioParticles™ products. Find out about these products and their applications by reviewing Probes for Following Receptor Binding and Phagocytosis—Section 16.1 in the Molecular Probes™ Handbook.

For pH-sensitive endocytosis assays, see our fluorescent pHrodo™ BioParticles™ conjugates.

For Research Use Only. Not for human or animal therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodFluorescence
Dye TypeUnconjugated
FormSolid
Quantity100 mg
Shipping ConditionRoom Temperature
SpeciesS. cerevisiae
For Use With (Equipment)Fluorescence Microscope
Product LineBioParticles
Product TypeBioparticle Conjugate
Unit Size100 mg
Contents & Storage
Store in freezer (-5 to -30°C).

Frequently asked questions (FAQs)

Are the Invitrogen BioParticles products sterile?

While the bacteria have been attenuated with formaldehyde and alcohol desiccation, the BioParticles products are not considered sterile, and we do not recommend incubation of more than 4 hours. This applies to all of our dye-labeled (pHrodo, Alexa Fluor, etc.) and unlabeled BioParticles products.

What is the type of bond that attaches the dyes to the BioParticles probes?

We use amine-reactive dyes to covalently attach fluorescent dyes to all of our BioParticles probes such as the Escherichia coli (K-12 strain) BioParticles probes, Staphylococcus aureus (Wood strain without protein A) BioParticles, and the Zymosan A (S. cerevisiae) BioParticles probes.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

What can the BioParticles product line be used for?

Fluorescent Bioparticles have been employed to study phagocytosis by fluorescence microscopy, quantitative spectrofluorometry, and flow cytometry. We offer E.Coli, S. aureus, and zymosan BioParticles conjugates covalently labeled with a variety of different fluorophores.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.