What are ZOOM Focusing buffers?
ZOOM Focusing Buffers are proprietary buffer formulations that facilitate optimal fractionation of proteins especially when using the basic ZOOM Disks. We offer ZOOM Focusing Buffer pH 3-7. ZOOM Focusing Buffer pH 7-12 has been discontinued.
ZOOM Focusing Buffers overcome the background streaking issues associated with the traditional method of using pH 3-10 ampholytes, thus ensuring optimal sample resolution.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
ZOOM Focusing Buffers overcome the background streaking issues associated with the traditional method of using pH 3-10 ampholytes, thus ensuring optimal sample resolution.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.