LentiArray™ CRISPR Positive Control Lentivirus, human HPRT, with GFP
Invitrogen™
LentiArray™ CRISPR Positive Control Lentivirus, human HPRT, with GFP
Optimizing delivery conditions is a critical step in any experiment utilizing the CRISPR/Cas9 system. The Invitrogen™ LentiArray CRISPR Positive ControlRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A328301 mL
A32060100 μL
Catalog number A32830
Price (USD)
2,051.65
Online exclusive
2,255.00
Save 203.35
Each
Quantity:
1 mL
Price (USD)
2,051.65
Online exclusive
2,255.00
Save 203.35
Each
Optimizing delivery conditions is a critical step in any experiment utilizing the CRISPR/Cas9 system. The Invitrogen™ LentiArray CRISPR Positive Control Lentivirus gRNA against HPRT is a highly efficient gRNA validated in multiple human cell lines that has achieved editing efficiencies of greater than 80%. It can be used to rapidly determine ideal delivery conditions in order to achieve maximum editing efficiency in your cell model of choice. This construct also expresses emGFP to provide a visual readout of successful transduction that can be used in determining Multiplicity of Infection (MOI).
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Delivery TypeLentiviral
For Use With (Application)Genome Editing
Product LineLentiArray
Product TypegRNA
PromoterU6
Quantity1 mL
Shipping ConditionDry Ice
Control TypePositive Control
FormatTube
RNAi TypegRNA
SpeciesHuman
Unit SizeEach
Contents & Storage
Store in ultra-cold freezer (-68°C to -85°C).