Product Image
Applied Biosystems™
QuantStudio™ 3 Real-Time PCR System, MiniAmp™ Plus Thermal Cycler, and Qubit Fluorometer Package
This is a package consisting of a QuantStudio 3 Real-Time PCR System with laptop, MiniAmp Plus Thermal Cycler, Qubit 4Read more
Have Questions?
Catalog NumberQuantity
A40479Package Solution
Catalog number A40479
Price (USD)
-
Quantity:
Package Solution
This is a package consisting of a QuantStudio 3 Real-Time PCR System with laptop, MiniAmp Plus Thermal Cycler, Qubit 4 Quantitation Starter Kit (including fluorometer), and reaction plates/films.
QuantStudio 3 Real-Time PCR System
The Applied Biosystems QuantStudio 3 Real-Time PCR System is designed for users who need an affordable, easy-to-use real-time PCR system that doesn't compromise performance and quality. The simplified Design and Analysis software is ideal for both first-time and experienced users. When connected to the Thermo Fisher Cloud, the QuantStudio 3 system provides access to your data wherever and whenever you want. Using proven OptiFlex technology (featuring four coupled channels and white LED) and featuring three independent Veriflex temperature zones, the QuantStudio 3 system enables improved data accuracy and sensitivity for a broad range of genomic applications.
Learn more about the QuantStudio 3 system >
MiniAmp Plus Thermal Cycler
The Applied Biosystems MiniAmp Plus Thermal Cycler is a compact, accurate, and economical thermal cycler designed to fit your lab’s basic thermal cycling needs. Innovations like a responsive color touch-screen and VeriFlex Blocks temperature-control technology enable simple, accurate optimization. The MiniAmp Plus Thermal Cycler can be accessed through the Thermo Fisher Cloud, giving you the freedom to design and securely upload your methods from any mobile device or desktop computer.
Learn more about the MiniAmp Plus Thermal Cycler ›
Qubit 4 Quantitation Starter Kit
The Qubit 4 Fluorometer is the latest version of the popular benchtop fluorometer designed to accurately measure DNA, RNA, and protein quantity. It also easily measures RNA integrity and quality. The easy-to-use touch screen menus make it easy to select and run the assays you need, with results displayed in just a few seconds. The Qubit 4 Quantitation Starter Kit includes:
• One Qubit 4 Fluorometer (Cat. No. Q33226)
• Qubit Assay Tubes (set of 500) (Cat. No. Q32856)
• Qubit 1X dsDNA HS Assay Kit, 100 assays
• Qubit dsDNA BR Assay Kit, 100 assays (Cat. No. Q32850)
• Qubit RNA IQ Assay Kit, 75 assays (Cat. No. Q33221)
• Qubit RNA HS Assay Kit, 100 assays (Cat. No. Q32852)
• Qubit RNA XR Assay Kit, 100 assays (Cat. No. Q33223)
• Qubit Protein Assay Kit, 100 assays (Cat. No. Q33211)
Learn more about the Qubit 4 Fluorometer and this starter kit ›
MicroAmp Optical 96-Well Reaction Plate with Barcode & Optical Adhesive Films
The Applied Biosystems MicroAmp Optical 96-Well Reaction Plate is optimized to provide unmatched temperature accuracy and uniformity for fast, efficient PCR amplification. This plate, constructed from a single rigid piece of polypropylene in a 96-well format, is compatible with Applied Biosystems 96-well real-time PCR systems and thermal cyclers.
The included MicroAmp Optical Adhesive Films reduce the chance of well-to-well contamination and sample evaporation when applied to the microplates.
Learn more about these plates and films ›
Specifications
For Use With (Application)PCR
Product LineApplied Biosystems
QuantityPackage Solution
Shipping ConditionApproved for Shipment at Room Temperature or on Wet Ice
Capacity96-well Plate
Product TypeThermal Cycler
Unit SizeEach
Package Components
MiniAmp™ Plus Thermal Cycler
Catalog Number
A37835
Unit Size
1 instrument
Qty
1
Qubit™ 4 Quantitation Starter Kit
Catalog Number
Q33227
Unit Size
1 kit
Qty
1
SMARTSTART ORNTATION,HALFDAY
Catalog Number
A28806
Unit Size
1 each
Qty
1
Contents & Storage
• QuantStudio 3 Real Time PCR System, 96-well, 0.2 mL, with laptop
• MiniAmp Plus Thermal Cycler
• MicroAmp Optical 96-well Plates w/Barcode & Optical Adhesive Films
• Qubit 4 Quantitation Starter Kit
Frequently asked questions (FAQs)
Can I downgrade the firmware on my QuantStudio 5?
You can perform this action if you have access to an Admin account on the QuantStudio instrument and if it is compatible for the downgrade (newer instruments are not compatible with v1.3.3). If your instrument serial number is below the following, then it can be downgraded:
QuantStudio 5 96-well, 0.1 mL block - SN 272514140
QuantStudio 5 96-well, 0.2 mL block - SN 2725210507
QuantStudio 5 384 well block - SN 272533285
Please also check if the instrument is being used with any validated protocols that may require a particular firmware version to be compliant. If you need the firmware itself, you can download it here.
The instructions to downgrade the firmware are on p.4 of the Release Notes: QuantStudioTM 3 & 5 Touchscreen Instrument Operating Software Version 1.3.3. It's the same process as an upgrade.
What is the admin pin for QuantStudio 3 and 5 Real-Time PCR Systems?
If the admin PIN has not been changed, 1234 should work. If it does not work, you can also try 0000.
However, if the admin PIN has been changed and none of the above PINs work, follow the instructions in the QuantStudio 3 and 5 Real-Time PCR Systems manual, page 100.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
Can I import a standard curve for my real-time PCR data into the Design & Analysis Software (v2.x)?
It is possible to import a standard curve into the Design & Analysis Software (v2.x) if you are analyzing results from one of the following Applied Biosystems real-time PCR instruments:
• 7900HT Fast Real-Time PCR System
• 7500/7500 Fast Real-Time PCR System
• StepOne/StepOne Plus Real-Time PCR System
• ViiA 7 Real Time PCR System
• QuantStudio 1 Real-Time PCR System
• QuantStudio 3 & 5 Real-Time PCR System
• QuantStudio 6 & 7 Flex Real-Time PCR System
• QuantStudio 6 & 7 Pro Real-Time PCR Systems
• QuantStudio 12K Flex Real-Time PCR system
Follow the instructions below to import a standard curve for analysis in the Design & Analysis Software (v2.x):
1. Open the data file in the Design & Analysis Software (v2.x).
2. Click on Actions.
3. Click on "Standard Curve Analysis Setting".
4. Select "External Standard Curves" and click on "Import".
5. Browse to the proper .csv file and click "Open".
6. Click "Apply".
Can I import a standard curve for my real-time PCR data into the QuantStudio Design & Analysis Software (v1.x)?
If you are analyzing results from the QuantStudio 1 Real-Time PCR System, QuantStudio 3 Real-Time PCR System, or QuantStudio 5 Real-Time PCR System, it is possible to import a standard curve into the QuantStudio Design & Analysis Software (v1.x) following the instructions below:
1. Open the data file in the QuantStudio Design & Analysis Software (v1.x).
2. Go to Analysis Settings.
3. Click on the "Standard Curve Settings" tab.
4. Select the option to "Use a standard curve imported from another experiment" and click on "Import".
5. Browse to and highlight the proper data file and click "Select".
6. Click Apply.