Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001039960.2 | NP_001035049.1 | 8 | 1107 | |
NM_001258401.2 | NP_001245330.1 | 8 | 1053 | ||
NM_001258402.1 | NP_001245331.1 | 8 | 1107 | ||
NM_001258403.1 | NP_001245332.1 | 8 | 1179 | ||
NM_001267615.1 | NP_001254544.1 | 8 | 1053 | ||
XM_006719700.1 | XP_006719763.1 | 8 | 1108 | ||
XM_011539010.1 | XP_011537312.1 | 8 | 1108 | ||
XM_011539011.1 | XP_011537313.1 | 8 | 1053 | ||
XM_017020240.1 | XP_016875729.1 | 8 | 1135 | ||
XM_017020241.1 | XP_016875730.1 | 8 | 1467 | ||
XM_017020242.1 | XP_016875731.1 | 8 | 1122 | ||
GenBank mRNA | AB018282.1 | - | 10 | 1187 | |
AF069512.1 | - | 8 | 944 | ||
AF107099.1 | - | 8 | 943 | ||
AK128321.1 | - | 9 | 1607 | ||
AK295315.1 | - | 8 | 1053 | ||
AK299545.1 | - | 5 | 739 | ||
AL831915.1 | - | 8 | 1179 | ||
BC025994.1 | - | 8 | 995 | 4822306 | |
DQ063579.2 | - | 8 | 1096 | ||
DQ975204.1 | - | 8 | 923 | ||
DQ996398.1 | - | 8 | 1157 | ||
DQ996537.1 | - | 8 | 923 |