Gene Transcripts

Gene Symbol
TGM5
Entrez Gene ID
9333
Gene Name
transglutaminase 5
Gene Aliases
PSS2, TGASE5, TGASEX, TGM6, TGMX, TGX
Chromosome Location
Chr. 15: 43232595 - 43266857 on Build GRCh38
UniGene ID
Hs.129719
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004245.3 NP_004236.1 2 173
NM_201631.3 NP_963925.2 2 173
GenBank mRNA AF035960.1 - 2 173
AF035961.1 - 2 173
BC119009.1 - 2 173 40085374
BC122859.1 - 2 173 40085372

View Full Product Details