Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001039960.2 | NP_001035049.1 | 2 | 264 | |
NM_001258401.2 | NP_001245330.1 | 2 | 210 | ||
NM_001258402.1 | NP_001245331.1 | 2 | 264 | ||
NM_001258403.1 | NP_001245332.1 | 2 | 336 | ||
NM_001267615.1 | NP_001254544.1 | 2 | 210 | ||
NR_047672.1 | 2 | 264 | |||
XM_006719700.1 | XP_006719763.1 | 2 | 265 | ||
XM_011539010.1 | XP_011537312.1 | 2 | 265 | ||
XM_011539011.1 | XP_011537313.1 | 2 | 210 | ||
XM_017020240.1 | XP_016875729.1 | 2 | 273 | ||
XM_017020241.1 | XP_016875730.1 | 2 | 624 | ||
XM_017020242.1 | XP_016875731.1 | 2 | 279 | ||
GenBank mRNA | AB018282.1 | - | 4 | 344 | |
AF069512.1 | - | 2 | 101 | ||
AF107099.1 | - | 2 | 100 | ||
AK128321.1 | - | 3 | 530 | ||
AK295315.1 | - | 2 | 210 | ||
AL831915.1 | - | 2 | 336 | ||
BC025994.1 | - | 2 | 152 | 4822306 | |
DA521806.1 | - | 2 | 265 | ||
DB220099.1 | - | 2 | 264 | ||
DQ063579.2 | - | 2 | 253 | ||
DQ975204.1 | - | 2 | 80 | ||
DQ996398.1 | - | 2 | 80 | ||
DQ996537.1 | - | 2 | 80 |