Gene Transcripts

Gene Symbol
GATA4
Entrez Gene ID
2626
Gene Name
GATA binding protein 4
Gene Aliases
ASD2, TACHD, TOF, VSD1
Chromosome Location
Chr. 8: 11676919 - 11760002 on Build GRCh38
UniGene ID
Hs.243987
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001308093.1 NP_001295022.1 2 1166
NM_001308094.1 NP_001295023.1 3 506
NM_002052.4 NP_002043.2 3 1317
XM_005272385.4 XP_005272442.1 4 1389
XM_005272386.1 XP_005272443.1 4 1577
XM_006716248.1 XP_006716311.1 4 1498
XM_011543817.2 XP_011542119.1 4 1568
XM_011543818.2 XP_011542120.1 3 1706
XM_017013312.1 XP_016868801.1 3 2022
GenBank mRNA AK097060.1 - 3 466
AY740706.1 - 2 709
BC101580.1 - 2 786 8069086
BC105108.1 - 2 784 8144111
BC143434.1 - 2 786 9051942
BC143479.1 - 2 789 9051987
D78260.1 - 2 998
DA900813.1 - 2 247
L34357.1 - 2 946

View Full Product Details