Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001286102.1 | NP_001273031.1 | 9 | 1947 | |
NM_003269.4 | NP_003260.1 | 9 | 2540 | ||
GenBank mRNA | AF220532.1 | - | 9 | 2539 | |
AK131541.1 | - | 9 | 1947 | ||
BC028031.1 | - | 9 | 2037 | 5242079 |