Gene Transcripts

Gene Symbol
Gls
Entrez Gene ID
24398
Gene Name
glutaminase
Gene Aliases
Glut, RATGLUT
Chromosome Location
Chr. 9: 54212593 - 54284879 on Build Rnor_6.0
UniGene ID
Rn.5762
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109968.1 NP_001103438.1 2 522
NM_012569.2 NP_036701.2 2 522
XM_008767075.1 XP_008765297.1 2 693
GenBank mRNA M65150.1 - 2 529

View Full Product Details