Gene Transcripts
- Gene Symbol
- Orm1
- Entrez Gene ID
- 24614
- Gene Name
- orosomucoid 1
- Gene Aliases
- AGP, Agpa1, OMD, Orm
- Chromosome Location
- Chr. 5: 79179668 - 79182820 on Build Rnor_6.0
- UniGene ID
- Rn.10295
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_053288.2 |
NP_445740.1
|
3, 4 |
377 |
|
GenBank mRNA |
FQ209739.1 |
- |
3, 4 |
379 |
|
FQ210338.1 |
- |
3, 4 |
379 |
|
FQ218200.1 |
- |
3, 4 |
382 |
|
FQ218302.1 |
- |
3, 4 |
379 |
|
FQ219945.1 |
- |
3, 4 |
383 |
|
FQ220896.1 |
- |
3, 4 |
383 |
|
FQ221011.1 |
- |
3, 4 |
382 |
|
FQ221095.1 |
- |
3, 4 |
378 |
|
FQ221803.1 |
- |
2, 3 |
173 |
|
FQ221848.1 |
- |
3, 4 |
379 |
|
FQ222215.1 |
- |
3, 4 |
379 |
|
FQ222231.1 |
- |
2, 3 |
190 |
|
FQ222829.1 |
- |
3, 4 |
379 |
|
FQ223035.1 |
- |
3, 4 |
379 |
|
FQ228371.1 |
- |
3, 4 |
383 |
|
FQ228464.1 |
- |
3, 4 |
377 |
|
FQ228466.1 |
- |
3, 4 |
379 |
|
FQ228489.1 |
- |
3, 4 |
379 |
|
FQ228584.1 |
- |
3, 4 |
379 |
|
FQ228800.1 |
- |
3, 4 |
379 |
|
FQ229026.1 |
- |
3, 4 |
378 |
|
FQ229053.1 |
- |
2, 3 |
103 |
|
FQ229515.1 |
- |
3, 4 |
297 |
|
FQ229654.1 |
- |
3, 4 |
377 |
|
FQ230009.1 |
- |
3, 4 |
383 |
|
J00696.1 |
- |
3, 4 |
378 |
|
V01216.1 |
- |
3, 4 |
377 |
|
View Full Product Details