Gene Transcripts

Gene Symbol
Slc15a4
Entrez Gene ID
246280
Gene Name
solute carrier family 15 (oligopeptide transporter), member 4
Gene Aliases
Chromosome Location
Chr. 12: 32753824 - 32773277 on Build Rnor_6.0
UniGene ID
Rn.10770
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144758.1 NP_653359.1 1 550
GenBank mRNA AB000280.1 - 1 550
BC087709.1 - 1 549 7315369

View Full Product Details