Gene Transcripts
- Gene Symbol
- Hexa
- Entrez Gene ID
- 15211
- Gene Name
- hexosaminidase A
- Gene Aliases
- Hex-1
- Chromosome Location
- Chr. 9: 59539545 - 59565109 on Build GRCm38
- UniGene ID
- Mm.2284
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_010421.4 |
NP_034551.2
|
3, 4 |
477 |
|
GenBank mRNA |
AK075895.1 |
- |
3, 4 |
453 |
|
AK075911.1 |
- |
3, 4 |
478 |
|
AK080777.1 |
- |
3, 4 |
450 |
|
AK144168.1 |
- |
3, 4 |
605 |
|
AK150444.1 |
- |
3, 4 |
448 |
|
AK151960.1 |
- |
3, 4 |
452 |
|
AK159091.1 |
- |
3, 4 |
469 |
|
AK159814.1 |
- |
3, 4 |
463 |
|
AK159890.1 |
- |
3, 4 |
450 |
|
AK160112.1 |
- |
3, 4 |
437 |
|
AK167013.1 |
- |
3, 4 |
463 |
|
AK168186.1 |
- |
3, 4 |
477 |
|
AK170903.1 |
- |
3, 4 |
463 |
|
BC010755.1 |
- |
3, 4 |
446 |
4225233 |
U07631.1 |
- |
3, 4 |
469 |
|
X64331.1 |
- |
3, 4 |
420 |
|
View Full Product Details