Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006437.3 | NP_006428.2 | 34 | 5284 | |
XM_011534931.1 | XP_011533233.1 | 33 | 5334 | ||
XM_011534932.2 | XP_011533234.1 | 34 | 5501 | ||
GenBank mRNA | AF057160.1 | - | 34 | 5230 | |
AF158255.1 | - | 34 | 5284 | ||
D79999.2 | - | 33 | 5183 |