Gene Transcripts

Gene Symbol
Chordc1
Entrez Gene ID
66917
Gene Name
cysteine and histidine-rich domain (CHORD)-containing, zinc-binding protein 1
Gene Aliases
1110001O09Rik, AA409036, Chp-1, morgana
Chromosome Location
Chr. 9: 18292233 - 18314000 on Build GRCm38
UniGene ID
Mm.103534
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025844.2 NP_080120.1 2, 3 252
GenBank mRNA AK003259.1 - 2, 3 253
AK012747.1 - 2, 3 237
AK132696.1 - 232
BC018374.1 - 2, 3 252 4236535

View Full Product Details