Gene Transcripts

Gene Symbol
Dnajc19
Entrez Gene ID
67713
Gene Name
DnaJ (Hsp40) homolog, subfamily C, member 19
Gene Aliases
1810055D05Rik, AA959924, Tim14
Chromosome Location
Chr. 3: 34057280 - 34081354 on Build GRCm38
UniGene ID
Mm.274266
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001026211.2 NP_001021382.1 4 328
NM_001286972.1 NP_001273901.1 4 328
NM_001286973.1 NP_001273902.1 3 276
NM_026332.4 NP_080608.3 4 328
GenBank mRNA AK007866.1 - 4 296
AK008267.1 - 4 272
AK010204.1 - 4 278
AK012051.1 - 4 280
AK076602.1 - 287
BC020327.1 - 4 261
BC024335.1 - 4 236 4974988
BY149679.1 - 4 301
CA469052.1 - 4 365
CJ123658.1 - 3 233
EL962106.1 - 4 274

View Full Product Details