Gene Transcripts

Gene Symbol
Mapk1ip1
Entrez Gene ID
69546
Gene Name
mitogen-activated protein kinase 1 interacting protein 1
Gene Aliases
2310009E07Rik, AU043776, MISS
Chromosome Location
Chr. 7: 138835818 - 138846267 on Build GRCm38
UniGene ID
Mm.31598
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001045483.1 NP_001038948.1 2 1431
NM_027115.2 NP_081391.1 2 1327
GenBank mRNA AK009250.1 - 2 1318
BC048375.1 - 2 1424
BC055699.1 - 3 1795
BC069850.1 - 1 995 5366053

View Full Product Details